ContactUsLinkComponent
Comfort rack for chest freezers for 14 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 592x140x140mm 1 * 1 items
Catalog # TENATE24442
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 14 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 592x140x140mm 1 * 1 items
Catalog # TENATE24442
Supplier:  TENAK