ContactUsLinkComponent
Comfort rack for chest freezers for 13 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 550x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24440B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 13 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 550x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24440B
Supplier:  TENAK