ContactUsLinkComponent
Comfort rack for chest freezers for 10 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 424x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24434B10
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 10 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 424x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24434B10
Supplier:  TENAK