ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 3x5=15 cryoboxes, 75mm high, stainless steel, sliding shelves, dimensions (HxDxW) 239x68 3x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24286B10
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 3x5=15 cryoboxes, 75mm high, stainless steel, sliding shelves, dimensions (HxDxW) 239x68 3x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24286B10
Supplier:  TENAK