ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 2x3=6 cryoboxes, 75mm high, stainless steel, sliding shelves, dimensions (HxDxW) 160x424 x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24262B10
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 2x3=6 cryoboxes, 75mm high, stainless steel, sliding shelves, dimensions (HxDxW) 160x424 x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24262B10
Supplier:  TENAK