ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 3x2=6 cryoboxes, 50mm high, stainless steel, sliding shelves, dimensions (HxDxW) 167x287 x140mm 1 * 1 items
Catalog # TENATE24216
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 3x2=6 cryoboxes, 50mm high, stainless steel, sliding shelves, dimensions (HxDxW) 167x287 x140mm 1 * 1 items
Catalog # TENATE24216
Supplier:  TENAK