ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 6x5=30 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 217x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24152B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 6x5=30 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 217x68 3x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24152B
Supplier:  TENAK