ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 10x4=40 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 361x5 62x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE24140B
 :  TENAK
New Product
Comfort rack for upright freezers for (HxD) 10x4=40 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 361x5 62x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE24140B
 :  TENAK