ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 8x4=32 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 289x56 2x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24136B10
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 8x4=32 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 289x56 2x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24136B10
Supplier:  TENAK