ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 5x3=15 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 181x42 4x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24120B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 5x3=15 cryoboxes, 32mm high, stainless steel, sliding shelves, dimensions (HxDxW) 181x42 4x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24120B
Supplier:  TENAK