ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 8x5=40 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 341x68 3x140mm 1 * 1 items
Catalog # TENATE24042
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 8x5=40 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 341x68 3x140mm 1 * 1 items
Catalog # TENATE24042
Supplier:  TENAK