ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 9x4=36 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 384x56 2x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24032B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 9x4=36 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 384x56 2x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24032B
Supplier:  TENAK