ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 6x4=24 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 256x56 2x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24026B10
 :  TENAK
New Product
Comfort rack for upright freezers for (HxD) 6x4=24 cryoboxes, 40mm high, stainless steel, sliding shelves, dimensions (HxDxW) 256x56 2x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE24026B10
 :  TENAK