ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 2x4=8 cryoboxes, 75mm high, stainless steel, dimensions (HxDxW) 160x561x139mm, delivere d including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23180B10
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 2x4=8 cryoboxes, 75mm high, stainless steel, dimensions (HxDxW) 160x561x139mm, delivere d including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23180B10
Supplier:  TENAK