ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 6x5=30 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 217x683x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23082B10
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 6x5=30 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 217x683x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23082B10
Supplier:  TENAK