ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 9x4=36 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 325x561x139mm, deliver ed including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE23068B
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 9x4=36 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 325x561x139mm, deliver ed including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE23068B
Supplier:  TENAK