ContactUsLinkComponent
Classic rack for chest freezers for 6 cryoboxes, 100mm high, stainless steel, with locking rod, dimensions (HxDxW) 628x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21474B10
Supplier:  TENAK
New Product
Classic rack for chest freezers for 6 cryoboxes, 100mm high, stainless steel, with locking rod, dimensions (HxDxW) 628x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21474B10
Supplier:  TENAK