ContactUsLinkComponent
Classic rack for chest freezers for 5 cryoboxes, 100mm high, stainless steel, with locking rod, dimensions (HxDxW) 524x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE21472B10
 :  TENAK
New Product
Classic rack for chest freezers for 5 cryoboxes, 100mm high, stainless steel, with locking rod, dimensions (HxDxW) 524x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE21472B10
 :  TENAK