ContactUsLinkComponent
Classic rack for chest freezers for 5 cryoboxes, 50mm high, stainless steel, with locking rod, dimensions (HxDxW) 279x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21449B10
Supplier:  TENAK
New Product
Classic rack for chest freezers for 5 cryoboxes, 50mm high, stainless steel, with locking rod, dimensions (HxDxW) 279x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21449B10
Supplier:  TENAK