ContactUsLinkComponent
Side up rack for both chest and upright freezers for 5 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 140x695x36mm, deliv ered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE21392B
 :  TENAK
New Product
Side up rack for both chest and upright freezers for 5 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 140x695x36mm, deliv ered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
  TENATE21392B
 :  TENAK