ContactUsLinkComponent
Classic rack for chest freezers for 6 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 256x140x140mm 1 * 1 items
Catalog # TENATE21370
Supplier:  TENAK
New Product
Classic rack for chest freezers for 6 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 256x140x140mm 1 * 1 items
Catalog # TENATE21370
Supplier:  TENAK