ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 3x7=21 plates, 55mm high, aluminium, dimensions (HxDxW) 172x630x135mm 1 * 1 items
Catalog # TENATE15648
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 3x7=21 plates, 55mm high, aluminium, dimensions (HxDxW) 172x630x135mm 1 * 1 items
Catalog # TENATE15648
Supplier:  TENAK