ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 55mm high, aluminium, dimensions (HxDxW) 343x450x135mm 1 * 1 items
Catalog # TENATE15634
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 55mm high, aluminium, dimensions (HxDxW) 343x450x135mm 1 * 1 items
Catalog # TENATE15634
Supplier:  TENAK