ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 11x7=77 plates, 35mm high, aluminium, dimensions (HxDxW) 408x630x135mm 1 * 1 items
Catalog # TENATE15346
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 11x7=77 plates, 35mm high, aluminium, dimensions (HxDxW) 408x630x135mm 1 * 1 items
Catalog # TENATE15346
Supplier:  TENAK