ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 10x5=50 plates, 35mm high, aluminium, dimensions (HxDxW) 371x450x135mm 1 * 1 items
Catalog # TENATE15320
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 10x5=50 plates, 35mm high, aluminium, dimensions (HxDxW) 371x450x135mm 1 * 1 items
Catalog # TENATE15320
Supplier:  TENAK