ContactUsLinkComponent
Eco Alu MTP racks for chest freezers for 12 SBS format plates, 55mm high, aluminium, dimensions (HxDxW) 689x135x92mm 1 * 1 items
Catalog # TENATE15274
Supplier:  TENAK
New Product
Eco Alu MTP racks for chest freezers for 12 SBS format plates, 55mm high, aluminium, dimensions (HxDxW) 689x135x92mm 1 * 1 items
Catalog # TENATE15274
Supplier:  TENAK