ContactUsLinkComponent
Eco AluRack for chest freezers for 5 cryoboxes, 130mm high, aluminium, dimensions (HxDxW) 676x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11576B10
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 5 cryoboxes, 130mm high, aluminium, dimensions (HxDxW) 676x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11576B10
Supplier:  TENAK