ContactUsLinkComponent
Eco AluRack for chest freezers for 6 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 485x140x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11538B10
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 6 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 485x140x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11538B10
Supplier:  TENAK