ContactUsLinkComponent
Eco AluRack for chest freezers for 15 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 641x140x140mm, delivered including standar d white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11444B
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 15 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 641x140x140mm, delivered including standar d white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11444B
Supplier:  TENAK