ContactUsLinkComponent
Eco AluRack for chest freezers for 13 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 557x140x140mm, delivered including standar d white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11440B
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 13 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 557x140x140mm, delivered including standar d white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11440B
Supplier:  TENAK