ContactUsLinkComponent
Eco AluRack for chest freezers for 10 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 431x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11434B10
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 10 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 431x140x140mm, delivered including standar d white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11434B10
Supplier:  TENAK