ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 3x4=12 cryoboxes, 130mm high, aluminium, dimensions (HxDxW) 401x564x139mm, delivered incl uding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11348B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 3x4=12 cryoboxes, 130mm high, aluminium, dimensions (HxDxW) 401x564x139mm, delivered incl uding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11348B10
Supplier:  TENAK