ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 3x3=9 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 240x425x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11264B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 3x3=9 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 240x425x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11264B10
Supplier:  TENAK