ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 3x3=9 cryoboxes, 50mm high, aluminium, dimensions (HxDxW) 168x425x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Référence TENATE11220B10
Fournisseur:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 3x3=9 cryoboxes, 50mm high, aluminium, dimensions (HxDxW) 168x425x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Référence TENATE11220B10
Fournisseur:  TENAK