ContactUsLinkComponent
Disposable Vacuum Box 90L Includes built-in handles Vacuum insulated carton box designed for long therm transports inner dimensions: 560x365x435 mm outer dimensions: 640x445x530 mm weight: 11 kg fits for: Accu 3L, Accu 2L and Accu 1L 1 * 1 SET
  DLTTB120004_G
 :  DELTA T
New Product
Disposable Vacuum Box 90L Includes built-in handles Vacuum insulated carton box designed for long therm transports inner dimensions: 560x365x435 mm outer dimensions: 640x445x530 mm weight: 11 kg fits for: Accu 3L, Accu 2L and Accu 1L 1 * 1 SET
  DLTTB120004_G
 :  DELTA T