ContactUsLinkComponent
ASM-IR behrotest analysis unit Kjeldahl for 12 samples, with distillation unit, fume eva- ecuator, burette, mag. stirrer consisting of: -digestion unit Inkjel1225P, for 12 x 250 ml - scrubber behrosog3 - distillation unit S3 - titration station STI 1 * 1 items
Catalog # BEHRB00637680
Supplier:  BEHR
New Product
ASM-IR behrotest analysis unit Kjeldahl for 12 samples, with distillation unit, fume eva- ecuator, burette, mag. stirrer consisting of: -digestion unit Inkjel1225P, for 12 x 250 ml - scrubber behrosog3 - distillation unit S3 - titration station STI 1 * 1 items
Catalog # BEHRB00637680
Supplier:  BEHR