ContactUsLinkComponent
High Precision Bench Scale SCK: PBK9890R5W3R305200 Commodity Code: 84238220000 Country of Origin: DE Base Model: PBK989 Application: Hazardous area Cat 3GD/Div2 Capacity: 30 kg Platform size: AB, 280x350mm IP-Protection: IP66/68 Resolution: 240.000d/ 1 * 1 items
Catalog # METB30214791-30KG
Supplier:  Mettler - Toledo
New Product
High Precision Bench Scale SCK: PBK9890R5W3R305200 Commodity Code: 84238220000 Country of Origin: DE Base Model: PBK989 Application: Hazardous area Cat 3GD/Div2 Capacity: 30 kg Platform size: AB, 280x350mm IP-Protection: IP66/68 Resolution: 240.000d/ 1 * 1 items
Catalog # METB30214791-30KG
Supplier:  Mettler - Toledo