ContactUsLinkComponent
PREFILTER ACRYLIC POLYESTER FLAT CELL, FRAME AND NETS IN GALVANIZED STEEL, G3/COARSE 40%, 495X295X20MM 1 * 1 ST
Catalog # MSPPXVLHG006
PREFILTER ACRYLIC POLYESTER FLAT CELL, FRAME AND NETS IN GALVANIZED STEEL, G3/COARSE 40%, 495X295X20MM 1 * 1 ST
Catalog # MSPPXVLHG006
Frequently Bought Together