The height of this ankle boot ensures the ankle is protected and supported. The upper comprises of a heavy duty textile, combined wi th a composite toecap, ESD properties and excellent slip resistance. A padded collar offers exceptional comfort. 1 * 1 items
Catalog # PRTWFE01BTE43
Supplier: Portwest