[2'-(AMINO-N)[1,1'-BIPHENYL]-2-YL-C][B 1 * 1 g
AMBDA830151-1G
: AMBEED
Frequently Bought Together
Barbituric acid, Supelco®
: Merck
Hexane (mixture of isomers) 99+% for HPLC
: Thermo Fisher Scientific
Glycerine 99+%, extra pure
: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
: Thermo Fisher Scientific
Tetrachloroethylene 99%, extra pure
: Thermo Fisher Scientific
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, Reagent Grade
: Honeywell Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
: PanReac AppliChem
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
: Honeywell Chemicals