DICHLOROBIS(TRICYCLOHEXYLPHOSPHINE)PALLA 1 * 10 g
Catalog # AMBDA113862-10G
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
n-Octane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Pyrene for synthesis, Sigma-Aldrich®
Supplier: Merck
Water, SupraSolv® for headspace gas chromatography, Supelco®
Supplier: Merck
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dichloroethane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.9% Reag. Ph. Eur., ACS
Supplier: PanReac AppliChem
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific