ContactUsLinkComponent
SIMPLE PLEX CARTRIDGE KIT FOR 32 SAMPLES, CONTAINING ASC, CASPASE-1 2ND GEN, IL-18/1L-1FA, IL-1RA/IL-1F3 FOR USE WITH HUMAN PLASMA/ SERUM. INCLUDES WASH BUFFER AND SAMPLE DILUENT SD13. 1 * 1 ST
Catalog # PRTSSPCKC-PS007360
Supplier:  PROTEIN SIMPLE
SIMPLE PLEX CARTRIDGE KIT FOR 32 SAMPLES, CONTAINING ASC, CASPASE-1 2ND GEN, IL-18/1L-1FA, IL-1RA/IL-1F3 FOR USE WITH HUMAN PLASMA/ SERUM. INCLUDES WASH BUFFER AND SAMPLE DILUENT SD13. 1 * 1 ST
Catalog # PRTSSPCKC-PS007360
Supplier:  PROTEIN SIMPLE
Frequently Bought Together