ContactUsLinkComponent
,,6 components: Bromides (Br-) 125mg/l Fluorides (F-) 125mg/l Nitrates (NO3-) 125mg/l Sulphates (SO42-) 500mg/l Chlorides (Cl-) 1250mg/l Phosphates (PO43-) 125mg/l in Water ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,, 1 * 100 mL
Catalog # CMPLRM115683L1
New Product
,,6 components: Bromides (Br-) 125mg/l Fluorides (F-) 125mg/l Nitrates (NO3-) 125mg/l Sulphates (SO42-) 500mg/l Chlorides (Cl-) 1250mg/l Phosphates (PO43-) 125mg/l in Water ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,, 1 * 100 mL
Catalog # CMPLRM115683L1