S3s hro ci hi lg fo sr ankle shoe, water resistant nabutek leather smellstop lining, fortrex puncture resistant patent + dry'n air footbed slimcap toe-cap fortrex sole pu + rubber 1 * 1 PAIR
Catalog # PRTWB1601BKO42
Supplier: Portwest