ContactUsLinkComponent
S3 hro ci src ankle shoe with overcap embedded in the sole, heel reinforcement and antitorsion system colour: black smellstop lin ing fresh'n flex puncture-resistant insole slimcap toe-cap pu double-density anti-fatigue airtech with hro outsole 1 * 1 PAIR
Catalog # PRTWB0888NBKD45
Supplier:  Portwest
New Product
S3 hro ci src ankle shoe with overcap embedded in the sole, heel reinforcement and antitorsion system colour: black smellstop lin ing fresh'n flex puncture-resistant insole slimcap toe-cap pu double-density anti-fatigue airtech with hro outsole 1 * 1 PAIR
Catalog # PRTWB0888NBKD45
Supplier:  Portwest