ContactUsLinkComponent
15 mL Vial Rack for Cleaning System Holds (36) 15 mL vials and closures PFA racks with PTFE post and handle Drip tray included Designed for use with vial cleaning system 550-01 May be used with some 4 L glass beakers (requires PFA beaker lid) 1 * 1 ST
Catalog # SAVI550-15
Supplier:  SAVILLEX
New Product
15 mL Vial Rack for Cleaning System Holds (36) 15 mL vials and closures PFA racks with PTFE post and handle Drip tray included Designed for use with vial cleaning system 550-01 May be used with some 4 L glass beakers (requires PFA beaker lid) 1 * 1 ST
Catalog # SAVI550-15
Supplier:  SAVILLEX