ContactUsLinkComponent
Azacitidine Formyl Amidine Analog (25 mg) (INTERNATIONAL COLD CHAIN SHIPMENT REQUIRED) (N-(Formyl amidino)-N'-Beta-D-ribofuranosylur ea) 1 * 25 mg
Catalog # 1A00260
Supplier:  USP
New Product
Azacitidine Formyl Amidine Analog (25 mg) (INTERNATIONAL COLD CHAIN SHIPMENT REQUIRED) (N-(Formyl amidino)-N'-Beta-D-ribofuranosylur ea) 1 * 25 mg
Catalog # 1A00260
Supplier:  USP