ContactUsLinkComponent
Cleanroom PE Tape 1154-3 white 3´ x 36 yards, Medium Tack, no residues, no metals, very low outgassing. Tears by hand, can write on with pen 16 Rolls/case 1 * 6 Roll
Catalog # UTAP1154WH300-P3D
Supplier:  ULTRATAPE
New Product
Cleanroom PE Tape 1154-3 white 3´ x 36 yards, Medium Tack, no residues, no metals, very low outgassing. Tears by hand, can write on with pen 16 Rolls/case 1 * 6 Roll
Catalog # UTAP1154WH300-P3D
Supplier:  ULTRATAPE