ContactUsLinkComponent
PROTECTION GLOVES (PAIR) JUGITEC PHARMA IN EPDM, CONFORM TO FDA POSITIVE LIST, ELECTRICAL CONDUCTIVE, BLACK COLOUR, AMBIDEXTROUS, LENGTH 920MM SIZE L, THK. 0,6MM, DIAM. 250MM 1 * 1 PAIR
Catalog # BSNC17BL06250920F
Supplier:  Basan
New Product
PROTECTION GLOVES (PAIR) JUGITEC PHARMA IN EPDM, CONFORM TO FDA POSITIVE LIST, ELECTRICAL CONDUCTIVE, BLACK COLOUR, AMBIDEXTROUS, LENGTH 920MM SIZE L, THK. 0,6MM, DIAM. 250MM 1 * 1 PAIR
Catalog # BSNC17BL06250920F
Supplier:  Basan