Transfer hose out of stainless steel with PTFE piping and phase separator 3/4ft, ength: 3 m 1 * 3 m
Catalog # KGWA2612-S4
Supplier: KGW
New Product
Transfer hose out of stainless steel with PTFE piping and phase separator 3/4ft, ength: 3 m 1 * 3 m
Catalog # KGWA2612-S4
Supplier: KGW
New Product
Frequently Bought Together
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for accu-jet® pro
Supplier: Brand
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
SONO-CHECK 1 * 30 items
Supplier: KING DISPOSABLES
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Cover gasket 1 * 1 items
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Accessories for Titrette®
Supplier: Brand
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Accessories for water purification millipore
Supplier: Merck
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Accessories for Rotavapor® R-3 / R-II / R-210 / R-215
Supplier: BUCHI